Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HES1
Entrez Gene ID
3280
Gene Name
hes family bHLH transcription factor 1
Gene Aliases
HES-1, HHL, HRY, bHLHb39
Chromosome Location
Chr.3: 194136142 - 194138612 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005524.3 NP_005515.1 1 - 2 351 100
GenBank mRNA AF264785.1 - 1 - 2 112 100
AK000415.1 - 1 - 2 348 100
AK300482.1 - 1 - 2 342 100
BC039152.1 - 1 - 2 299 100
CR541843.1 - 1 - 2 112 100

View Full Product Details