Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAS1L
Entrez Gene ID
81887
Gene Name
LAS1 like, ribosome biogenesis factor
Gene Aliases
Las1-like, dJ475B7.2
Chromosome Location
Chr.X: 65512582 - 65534806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170649.1 NP_001164120.1 1 - 2 327 59
NM_031206.4 NP_112483.1 1 - 2 327 59
XM_005262301.1 XP_005262358.1 1 - 2 312 59
XM_005262304.1 XP_005262361.1 1 - 2 312 59
XM_005262305.3 XP_005262362.1 1 - 2 312 59
XM_005262306.3 XP_005262363.1 1 - 2 312 59
XM_011531046.1 XP_011529348.1 1 - 2 312 59
XM_017029877.1 XP_016885366.1 1 - 2 312 59
XM_017029878.1 XP_016885367.1 1 - 2 312 59
XM_017029879.1 XP_016885368.1 1 - 2 312 59
XM_017029880.1 XP_016885369.1 1 - 2 312 59
GenBank mRNA AF116730.1 - 1 - 2 265 59
AK022587.1 - 1 - 2 277 59
AK074087.1 - 1 - 2 296 59
AK225059.1 - 1 - 2 260 59
AK295591.1 - 1 - 2 277 59
BC014545.1 - 1 - 2 311 59
BC018610.1 - 1 - 2 275 59
BC019302.1 - 1 - 2 272 59
DA619280.1 - 1 - 2 328 59

View Full Product Details