Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 10 - 11 1127 95
NM_001141974.2 NP_001135446.1 10 - 11 1127 95
NM_022089.3 NP_071372.1 10 - 11 1142 95
XM_005245810.1 XP_005245867.1 10 - 11 1139 95
XM_005245811.1 XP_005245868.1 10 - 11 1127 95
XM_005245812.1 XP_005245869.1 10 - 11 1115 95
XM_005245815.1 XP_005245872.1 10 - 11 1142 95
XM_006710512.1 XP_006710575.1 10 - 11 1124 95
XM_006710513.1 XP_006710576.1 10 - 11 1100 95
XM_011541128.1 XP_011539430.1 10 - 11 1142 95
XM_011541129.1 XP_011539431.1 10 - 11 1142 95
XM_017000844.1 XP_016856333.1 10 - 11 1142 95
XM_017000845.1 XP_016856334.1 10 - 11 1124 95
XM_017000846.1 XP_016856335.1 10 - 11 1100 95
XM_017000847.1 XP_016856336.1 10 - 11 1112 95
XM_017000848.1 XP_016856337.1 10 - 11 1142 95
XM_017000849.1 XP_016856338.1 10 - 11 1127 95
XM_017000850.1 XP_016856339.1 10 - 11 1142 95
GenBank mRNA AK290210.1 - 10 - 11 1083 95
AL354615.1 - 10 - 11 942 95
AY461712.1 - 10 - 11 941 95
BC030267.1 - 10 - 11 1070 95
BC034575.1 - 7 - 8 633 95

View Full Product Details