Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr.10: 76869601 - 77637819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014797.2 NP_001014797.1 13 - 14 1774 64
NM_001161352.1 NP_001154824.1 13 - 14 1774 64
NM_001161353.1 NP_001154825.1 13 - 14 1774 64
NM_001271518.1 NP_001258447.1 12 - 13 1612 64
NM_001271519.1 NP_001258448.1 13 - 14 1774 64
NM_001322829.1 NP_001309758.1 13 - 14 1774 64
NM_001322830.1 NP_001309759.1 13 - 14 1774 64
NM_001322832.1 NP_001309761.1 13 - 14 1774 64
NM_001322835.1 NP_001309764.1 13 - 14 1774 64
NM_001322836.1 NP_001309765.1 13 - 14 1774 64
NM_001322837.1 NP_001309766.1 13 - 14 1774 64
NM_001322838.1 NP_001309767.1 14 - 15 1734 64
NM_002247.3 NP_002238.2 13 - 14 1774 64
XM_005269776.3 XP_005269833.1 13 - 14 1763 64
XM_005269778.1 XP_005269835.1 13 - 14 1763 64
XM_005269781.2 XP_005269838.1 13 - 14 1695 64
XM_005269787.3 XP_005269844.1 13 - 14 1763 64
XM_005269789.2 XP_005269846.1 13 - 14 1695 64
XM_005269792.2 XP_005269849.1 13 - 14 1695 64
XM_005269796.2 XP_005269853.1 13 - 14 1695 64
XM_006717826.2 XP_006717889.1 13 - 14 1695 64
XM_011539773.1 XP_011538075.1 14 - 15 1895 64
XM_011539774.1 XP_011538076.1 14 - 15 1895 64
XM_011539775.1 XP_011538077.1 14 - 15 1895 64
XM_011539776.1 XP_011538078.1 14 - 15 1895 64
XM_011539777.1 XP_011538079.1 13 - 14 1763 64
XM_011539778.2 XP_011538080.1 13 - 14 1695 64
XM_011539779.1 XP_011538081.1 14 - 15 1895 64
XM_011539780.1 XP_011538082.1 13 - 14 1763 64
XM_011539781.2 XP_011538083.1 14 - 15 1895 64
XM_011539782.2 XP_011538084.1 14 - 15 1847 64
XM_011539783.1 XP_011538085.1 13 - 14 1763 64
XM_011539784.1 XP_011538086.1 14 - 15 1895 64
XM_011539785.1 XP_011538087.1 13 - 14 1763 64
XM_017016207.1 XP_016871696.1 14 - 15 1895 64
XM_017016208.1 XP_016871697.1 14 - 15 1895 64
XM_017016209.1 XP_016871698.1 14 - 15 1895 64
XM_017016210.1 XP_016871699.1 14 - 15 1895 64
XM_017016211.1 XP_016871700.1 14 - 15 1895 64
XM_017016212.1 XP_016871701.1 14 - 15 1895 64
XM_017016213.1 XP_016871702.1 14 - 15 1895 64
XM_017016214.1 XP_016871703.1 14 - 15 1895 64
XM_017016215.1 XP_016871704.1 13 - 14 1763 64
XM_017016216.1 XP_016871705.1 14 - 15 1895 64
XM_017016217.1 XP_016871706.1 14 - 15 1847 64
XM_017016218.1 XP_016871707.1 12 - 13 1220 64
XM_017016219.1 XP_016871708.1 14 - 15 1895 64
XM_017016222.1 XP_016871711.1 13 - 14 1763 64
XM_017016223.1 XP_016871712.1 11 - 12 1072 64
GenBank mRNA AB113382.1 - 13 - 14 1539 64
AB113575.1 - 13 - 14 1525 64
AB209488.1 - 12 - 13 1180 64
AB524033.1 - 12 - 13 1435 64
AF025999.1 - 13 - 14 1525 64
AF118141.1 - 13 - 14 1402 64
AK128392.1 - 4 - 5 2030 64
AK310379.1 - 14 - 15 1734 64
AY040849.1 - 13 - 14 1539 64
BC137115.1 - 13 - 14 1677 64
BC137137.1 - 13 - 14 1677 64
BC144496.1 - 13 - 14 1677 64
BX648925.1 - 8 - 9 756 64
U02632.1 - 6 - 7 632 64
U09384.1 - 13 - 14 1592 64
U11058.2 - 13 - 14 1488 64
U11717.1 - 13 - 14 1549 64
U13913.1 - 13 - 14 1675 64
U23767.1 - 13 - 14 1597 64

View Full Product Details