Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr.10: 76869601 - 77637819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014797.2 NP_001014797.1 23 - 24 2919 73
NM_001161352.1 NP_001154824.1 23 - 24 3081 73
NM_001161353.1 NP_001154825.1 23 - 24 3030 73
NM_001271518.1 NP_001258447.1 22 - 23 2757 73
NM_001271519.1 NP_001258448.1 23 - 24 2916 73
NM_001322829.1 NP_001309758.1 23 - 24 2916 73
NM_001322830.1 NP_001309759.1 24 - 25 2928 73
NM_001322832.1 NP_001309761.1 22 - 23 2907 73
NM_001322835.1 NP_001309764.1 23 - 24 2919 73
NM_001322836.1 NP_001309765.1 23 - 24 2916 73
NM_001322837.1 NP_001309766.1 23 - 24 2919 73
NM_001322838.1 NP_001309767.1 24 - 25 2954 73
NM_002247.3 NP_002238.2 22 - 23 2907 73
XM_005269776.3 XP_005269833.1 22 - 23 2896 73
XM_005269778.1 XP_005269835.1 24 - 25 3082 73
XM_005269781.2 XP_005269838.1 23 - 24 3002 73
XM_005269787.3 XP_005269844.1 22 - 23 2896 73
XM_005269789.2 XP_005269846.1 23 - 24 2840 73
XM_005269792.2 XP_005269849.1 23 - 24 2915 73
XM_005269796.2 XP_005269853.1 22 - 23 2828 73
XM_006717826.2 XP_006717889.1 22 - 23 2828 73
XM_011539773.1 XP_011538075.1 25 - 26 3214 73
XM_011539774.1 XP_011538076.1 25 - 26 3127 73
XM_011539775.1 XP_011538077.1 24 - 25 3202 73
XM_011539776.1 XP_011538078.1 24 - 25 3202 73
XM_011539777.1 XP_011538079.1 24 - 25 3082 73
XM_011539778.2 XP_011538080.1 23 - 24 3002 73
XM_011539779.1 XP_011538081.1 24 - 25 3115 73
XM_011539780.1 XP_011538082.1 24 - 25 3082 73
XM_011539781.2 XP_011538083.1 24 - 25 3040 73
XM_011539782.2 XP_011538084.1 23 - 24 2980 73
XM_011539783.1 XP_011538085.1 24 - 25 2995 73
XM_011539785.1 XP_011538087.1 23 - 24 2905 73
XM_017016207.1 XP_016871696.1 25 - 26 3214 73
XM_017016208.1 XP_016871697.1 25 - 26 3214 73
XM_017016209.1 XP_016871698.1 24 - 25 3202 73
XM_017016210.1 XP_016871699.1 25 - 26 3214 73
XM_017016211.1 XP_016871700.1 25 - 26 3127 73
XM_017016212.1 XP_016871701.1 24 - 25 3040 73
XM_017016213.1 XP_016871702.1 24 - 25 3115 73
XM_017016214.1 XP_016871703.1 23 - 24 3028 73
XM_017016215.1 XP_016871704.1 24 - 25 2995 73
XM_017016216.1 XP_016871705.1 24 - 25 3040 73
XM_017016217.1 XP_016871706.1 23 - 24 2980 73
XM_017016218.1 XP_016871707.1 22 - 23 2365 73
XM_017016222.1 XP_016871711.1 22 - 23 2896 73
XM_017016223.1 XP_016871712.1 21 - 22 2292 73
GenBank mRNA AB113382.1 - 23 - 24 2846 73
AB113575.1 - 23 - 24 2781 73
AB209488.1 - 22 - 23 2326 73
AB524033.1 - 22 - 23 2580 73
AF025999.1 - 22 - 23 2658 73
AF118141.1 - 22 - 23 2535 73
AK310379.1 - 24 - 25 2954 73
AY040849.1 - 22 - 23 2672 73
BC137115.1 - 22 - 23 2810 73
BC137137.1 - 22 - 23 2810 73
BC144496.1 - 23 - 24 2819 73
BX648925.1 - 19 - 20 2072 73
U02632.1 - 16 - 17 1852 73
U11058.2 - 22 - 23 2621 73
U11717.1 - 22 - 23 2682 73
U13913.1 - 22 - 23 2808 73
U23767.1 - 22 - 23 2730 73

View Full Product Details