Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT23
Entrez Gene ID
25984
Gene Name
keratin 23
Gene Aliases
CK23, HAIK1, K23
Chromosome Location
Chr.17: 40922696 - 40937643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015515.4 NP_056330.3 2 - 3 1046 143
GenBank mRNA AF102848.1 - 1 - 2 474 143
AK002047.1 - 1 - 2 458 143
AK225866.1 - 1 - 2 829 143
AK289449.1 - 1 - 2 541 143
AK310880.1 - 1 - 2 630 143
AK314604.1 - 1 - 2 258 143
BC028356.2 - 2 - 3 826 143
DA846104.1 - 1 - 2 258 143

View Full Product Details