The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024662.2 | NP_078938.2 | 3 - 4 | 447 | 109 |
GenBank mRNA | AB051496.1 | - | 3 - 4 | 368 | 109 |
AK001636.1 | - | 3 - 4 | 302 | 109 | |
AK022241.1 | - | 3 - 4 | 409 | 109 | |
AK074430.1 | - | - | 1025 | 109 | |
AK225341.1 | - | 3 - 4 | 409 | 109 | |
AL136882.1 | - | 3 - 4 | 304 | 109 | |
BC020205.1 | - | 3 - 4 | 269 | 109 | |
BC035558.1 | - | 3 - 4 | 381 | 109 | |
CN336530.1 | - | 3 - 4 | 449 | 109 |