The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000401.3 | NP_000392.3 | 6 - 7 | 1237 | 153 |
NM_207122.1 | NP_997005.1 | 6 - 7 | 1416 | 153 | |
XM_011519950.1 | XP_011518252.1 | 7 - 8 | 1235 | 153 | |
XM_011519951.1 | XP_011518253.1 | 7 - 8 | 1345 | 153 | |
GenBank mRNA | AK296713.1 | - | 6 - 7 | 930 | 153 |
AK312375.1 | - | 6 - 7 | 1293 | 153 | |
BC010058.1 | - | 6 - 7 | 1290 | 153 | |
BC068545.1 | - | 6 - 7 | 1256 | 153 | |
BX648142.1 | - | 6 - 7 | 1214 | 153 | |
U62740.1 | - | 6 - 7 | 1416 | 153 | |
U64511.1 | - | 5 - 6 | 1108 | 153 |