Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKTN
Entrez Gene ID
2218
Gene Name
fukutin
Gene Aliases
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
Chromosome Location
Chr.9: 105558117 - 105655950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006731.2 NP_006722.2 1 - 2 34 105
XM_006717014.2 XP_006717077.1 1 - 2 38 105
XM_011518374.2 XP_011516676.1 1 - 2 38 105
XM_011518381.2 XP_011516683.1 1 - 2 38 105
XM_017014474.1 XP_016869963.1 1 - 2 37 105
XM_017014475.1 XP_016869964.1 1 - 2 41 105
XM_017014479.1 XP_016869968.1 1 - 2 38 105
GenBank mRNA AK304460.1 - 1 - 2 32 105
AL601876.1 - 1 - 2 37 105

View Full Product Details