Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCIN
Entrez Gene ID
85477
Gene Name
scinderin
Gene Aliases
-
Chromosome Location
Chr.7: 12570349 - 12653603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112706.2 NP_001106177.1 14 - 15 2161 126
NM_033128.3 NP_149119.1 12 - 13 1486 126
GenBank mRNA AB067492.1 - 13 - 14 1983 126
AF276507.1 - 14 - 15 1951 126
AK027778.1 - 12 - 13 1486 126
AK075123.1 - 14 - 15 2020 126
AK075198.1 - 8 - 9 1002 126
AK290363.1 - 14 - 15 2054 126
BC021090.1 - 14 - 15 2000 126

View Full Product Details