Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCIN
Entrez Gene ID
85477
Gene Name
scinderin
Gene Aliases
-
Chromosome Location
Chr.7: 12570349 - 12653603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112706.2 NP_001106177.1 15 - 16 2228 118
NM_033128.3 NP_149119.1 13 - 14 1553 118
GenBank mRNA AB067492.1 - 14 - 15 2050 118
AF276507.1 - 15 - 16 2018 118
AK027778.1 - 13 - 14 1553 118
AK075123.1 - 15 - 16 2087 118
AK075198.1 - 9 - 10 1069 118
AK290363.1 - 15 - 16 2121 118
BC021090.1 - 15 - 16 2067 118

View Full Product Details