The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003185.3 | NP_003176.2 | 6 - 7 | 1975 | 128 |
GenBank mRNA | AK123937.1 | - | 2 - 3 | 824 | 128 |
U75308.1 | - | 8 - 9 | 1561 | 128 | |
Y11354.1 | - | 6 - 7 | 1969 | 128 |