Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
Chromosome Location
Chr.14: 55366391 - 55411858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014924.4 NP_055739.2 1 - 2 255 85
GenBank mRNA AK131251.1 - 1 - 2 255 85
AK292810.1 - 1 - 2 227 85
BC109089.1 - 1 - 2 240 85
BC109090.1 - 1 - 2 240 85

View Full Product Details