The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319139.1 | NP_001306068.1 | 5 - 6 | 627 | 65 |
NM_001319140.1 | NP_001306069.1 | 5 - 6 | 627 | 65 | |
NM_001319141.1 | NP_001306070.1 | 4 - 5 | 509 | 65 | |
NM_001319142.1 | NP_001306071.1 | 3 - 4 | 383 | 65 | |
NM_001625.3 | NP_001616.1 | 4 - 5 | 509 | 65 | |
NM_013411.4 | NP_037543.1 | 4 - 5 | 509 | 65 | |
GenBank mRNA | AB005621.1 | - | 4 - 5 | 464 | 65 |
AB005622.1 | - | 4 - 5 | 468 | 65 | |
AB451267.1 | - | 4 - 5 | 426 | 65 | |
AB451394.1 | - | 4 - 5 | 426 | 65 | |
AK291676.1 | - | 4 - 5 | 446 | 65 | |
AK295105.1 | - | 5 - 6 | 560 | 65 | |
AK307731.1 | - | 3 - 4 | 312 | 65 | |
AL557311.3 | - | 4 - 5 | 483 | 65 | |
AY080899.1 | - | 4 - 5 | 461 | 65 | |
AY080900.1 | - | 5 - 6 | 547 | 65 | |
BC009405.2 | - | 4 - 5 | 437 | 65 | |
BC026705.1 | - | 4 - 5 | 445 | 65 | |
BC070127.1 | - | 4 - 5 | 451 | 65 | |
BC090040.1 | - | 4 - 5 | 451 | 65 | |
BQ889289.1 | - | 5 - 6 | 593 | 65 | |
DC422465.1 | - | 4 - 5 | 509 | 65 | |
U39945.1 | - | 4 - 5 | 459 | 65 | |
U54645.1 | - | 4 - 5 | 429 | 65 | |
U84371.1 | - | 4 - 5 | 457 | 65 |