Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
Chromosome Location
Chr.7: 55964577 - 56000182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001483.2 NP_001474.1 4 - 5 410 73
GenBank mRNA AF029786.1 - 4 - 5 385 73
AJ001259.1 - 4 - 5 377 73
AK125036.1 - 1 - 2 2666 73
AK315648.1 - 4 - 5 395 73
BC000732.2 - 4 - 5 383 73
BC001837.2 - 4 - 5 383 73
BC030821.1 - 4 - 5 392 73
BT007112.1 - 4 - 5 377 73
CR407612.1 - 4 - 5 377 73
CX166452.1 - 4 - 5 410 73

View Full Product Details