Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA2
Entrez Gene ID
3908
Gene Name
laminin subunit alpha 2
Gene Aliases
LAMM
Chromosome Location
Chr.6: 128883141 - 129516566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000426.3 NP_000417.2 11 - 12 1719 73
NM_001079823.1 NP_001073291.1 11 - 12 1719 73
XM_005266981.3 XP_005267038.1 11 - 12 1719 73
XM_005266982.3 XP_005267039.1 11 - 12 1719 73
XM_011535820.2 XP_011534122.1 11 - 12 1719 73
XM_017010851.1 XP_016866340.1 11 - 12 4107 73
XM_017010853.1 XP_016866342.1 11 - 12 1719 73
GenBank mRNA Z26653.1 - 11 - 12 1663 73

View Full Product Details