Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA2
Entrez Gene ID
3908
Gene Name
laminin subunit alpha 2
Gene Aliases
LAMM
Chromosome Location
Chr.6: 128883141 - 129516566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000426.3 NP_000417.2 36 - 37 5339 135
NM_001079823.1 NP_001073291.1 36 - 37 5339 135
XM_005266981.3 XP_005267038.1 37 - 38 5603 135
XM_005266982.3 XP_005267039.1 37 - 38 5603 135
XM_011535820.2 XP_011534122.1 37 - 38 5603 135
XM_017010851.1 XP_016866340.1 37 - 38 7991 135
XM_017010852.1 XP_016866341.1 26 - 27 6880 135
XM_017010853.1 XP_016866342.1 37 - 38 5603 135
GenBank mRNA AB208922.1 - 11 - 12 1441 135
Z26653.1 - 36 - 37 5283 135

View Full Product Details