Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMA2
Entrez Gene ID
3908
Gene Name
laminin subunit alpha 2
Gene Aliases
LAMM
Chromosome Location
Chr.6: 128883141 - 129516566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000426.3 NP_000417.2 57 - 58 8187 123
NM_001079823.1 NP_001073291.1 56 - 57 8175 123
XM_005266981.3 XP_005267038.1 58 - 59 8451 123
XM_005266982.3 XP_005267039.1 57 - 58 8439 123
XM_011535820.2 XP_011534122.1 57 - 58 8445 123
XM_017010851.1 XP_016866340.1 58 - 59 10839 123
XM_017010852.1 XP_016866341.1 47 - 48 9728 123
GenBank mRNA AB208922.1 - 31 - 32 4277 123
M59832.1 - 17 - 18 2142 123
Z26653.1 - 57 - 58 8131 123

View Full Product Details