The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020964.2 | NP_066015.2 | 40 - 41 | 7111 | 59 |
XM_011526122.1 | XP_011524424.1 | 40 - 41 | 7111 | 59 | |
XM_017025889.1 | XP_016881378.1 | 40 - 41 | 7090 | 59 | |
XM_017025890.1 | XP_016881379.1 | 40 - 41 | 7093 | 59 | |
XM_017025891.1 | XP_016881380.1 | 39 - 40 | 6970 | 59 | |
XM_017025892.1 | XP_016881381.1 | 39 - 40 | 7136 | 59 | |
XM_017025893.1 | XP_016881382.1 | 32 - 33 | 4983 | 59 | |
GenBank mRNA | AK023817.1 | - | 24 - 25 | 3790 | 59 |
AL583915.1 | - | 1 - 2 | 120 | 59 | |
BC003695.1 | - | 4 - 5 | 790 | 59 | |
BC036911.1 | - | 4 - 5 | 797 | 59 | |
BC130614.1 | - | 23 - 24 | 3644 | 59 | |
BC144104.1 | - | 21 - 22 | 3305 | 59 | |
BC144105.1 | - | 22 - 23 | 3537 | 59 |