The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256119.1 | NP_001243048.1 | 6 - 7 | 676 | 66 |
NM_001256120.1 | NP_001243049.1 | 7 - 8 | 703 | 66 | |
NM_003491.3 | NP_003482.1 | 7 - 8 | 721 | 66 | |
GenBank mRNA | AY183134.1 | - | 6 - 7 | 782 | 66 |
AY183135.1 | - | 6 - 7 | 537 | 66 | |
BC000308.2 | - | 7 - 8 | 582 | 66 | |
BC019312.1 | - | 7 - 8 | 582 | 66 | |
BC063377.1 | - | 6 - 7 | 530 | 66 | |
BI192945.1 | - | 7 - 8 | 518 | 66 | |
CA313208.1 | - | 1 - 2 | 66 | ||
X77588.1 | - | 7 - 8 | 566 | 66 |