The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007118.3 | NP_009049.2 | 13 - 14 | 2412 | 97 |
NR_134469.1 | 13 - 14 | 2412 | 97 | ||
XM_011514107.2 | XP_011512409.1 | 13 - 14 | 2987 | 97 | |
XM_011514108.1 | XP_011512410.1 | 13 - 14 | 2286 | 97 | |
XM_011514109.2 | XP_011512411.1 | 13 - 14 | 6064 | 97 | |
XM_011514110.2 | XP_011512412.1 | 13 - 14 | 2522 | 97 | |
XM_017009801.1 | XP_016865290.1 | 13 - 14 | 2764 | 97 | |
XM_017009802.1 | XP_016865291.1 | 13 - 14 | 2764 | 97 | |
XM_017009803.1 | XP_016865292.1 | 6 - 7 | 1033 | 97 | |
GenBank mRNA | AB209754.1 | - | 8 - 9 | 1306 | 97 |
AK304786.1 | - | 13 - 14 | 2391 | 97 | |
BC035585.1 | - | 13 - 14 | 2337 | 97 |