Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIO
Entrez Gene ID
7204
Gene Name
trio Rho guanine nucleotide exchange factor
Gene Aliases
ARHGEF23, tgat
Chromosome Location
Chr.5: 14143350 - 14510204 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007118.3 NP_009049.2 14 - 15 2610 85
NR_134469.1 14 - 15 2610 85
XM_011514107.2 XP_011512409.1 14 - 15 3185 85
XM_011514108.1 XP_011512410.1 14 - 15 2484 85
XM_011514109.2 XP_011512411.1 14 - 15 6262 85
XM_011514110.2 XP_011512412.1 14 - 15 2720 85
XM_017009801.1 XP_016865290.1 14 - 15 2962 85
XM_017009802.1 XP_016865291.1 14 - 15 2962 85
XM_017009803.1 XP_016865292.1 7 - 8 1231 85
GenBank mRNA AB209754.1 - 9 - 10 1504 85
AF091395.1 - 14 - 15 2475 85
AK304786.1 - 14 - 15 2589 85
BC035585.1 - 14 - 15 2535 85
U42390.1 - 14 - 15 2475 85

View Full Product Details