Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC7A
Entrez Gene ID
64581
Gene Name
C-type lectin domain family 7 member A
Gene Aliases
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
Chromosome Location
Chr.12: 10116777 - 10130269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197948.2 NP_922939.1 4 - 5 671 73
NM_197949.2 NP_922940.1 3 - 4 533 73
GenBank mRNA AF400597.1 - 4 - 5 484 73
AF400598.1 - 3 - 4 346 73
AK297028.1 - 3 - 4 417 73
AY026771.1 - 3 - 4 346 73

View Full Product Details