Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
Chromosome Location
Chr.15: 49858238 - 50182096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024837.3 NP_079113.2 15 - 16 1591 76
NR_073597.1 15 - 16 1591 76
NR_073598.1 16 - 17 1698 76
XM_011522046.2 XP_011520348.1 17 - 18 1980 76
XM_011522047.2 XP_011520349.1 16 - 17 2505 76
XM_011522048.1 XP_011520350.1 16 - 17 1778 76
XM_011522049.2 XP_011520351.1 16 - 17 1888 76
XM_011522051.2 XP_011520353.1 17 - 18 1815 76
XM_011522052.2 XP_011520354.1 16 - 17 1873 76
XM_011522053.1 XP_011520355.1 16 - 17 1654 76
XM_011522056.2 XP_011520358.2 16 - 17 2040 76
XM_011522058.2 XP_011520360.1 12 - 13 2249 76
XM_011522059.1 XP_011520361.1 11 - 12 1270 76
XM_011522060.1 XP_011520362.1 14 - 15 1505 76
XM_011522061.1 XP_011520363.1 13 - 14 1420 76
XM_011522062.1 XP_011520364.1 11 - 12 1259 76
XM_011522063.1 XP_011520365.1 11 - 12 1241 76
XM_011522064.1 XP_011520366.1 7 - 8 942 76
XM_011522069.2 XP_011520371.1 6 - 7 763 76
XM_011522070.1 XP_011520372.1 2 - 3 183 76
XM_017022587.1 XP_016878076.1 15 - 16 1956 76
XM_017022588.1 XP_016878077.1 15 - 16 1881 76
XM_017022590.1 XP_016878079.1 17 - 18 1792 76
XM_017022591.1 XP_016878080.1 16 - 17 1739 76
XM_017022592.1 XP_016878081.1 15 - 16 1802 76
XM_017022593.1 XP_016878082.1 16 - 17 2040 76
XM_017022594.1 XP_016878083.1 12 - 13 1300 76
XM_017022595.1 XP_016878084.1 12 - 13 3305 76
XM_017022596.1 XP_016878085.1 7 - 8 933 76
XM_017022597.1 XP_016878086.1 16 - 17 2040 76
XM_017022598.1 XP_016878087.1 16 - 17 2040 76
XM_017022599.1 XP_016878088.1 16 - 17 2040 76
GenBank mRNA AB075819.1 - 16 - 17 1698 76
BC144297.1 - 15 - 16 1587 76

View Full Product Details