Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
Chromosome Location
Chr.15: 49858238 - 50182096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024837.3 NP_079113.2 2 - 3 165 94
NR_073596.1 2 - 3 165 94
NR_073597.1 2 - 3 165 94
NR_073598.1 2 - 3 184 94
XM_011522046.2 XP_011520348.1 3 - 4 470 94
XM_011522047.2 XP_011520349.1 2 - 3 995 94
XM_011522048.1 XP_011520350.1 2 - 3 268 94
XM_011522049.2 XP_011520351.1 2 - 3 378 94
XM_011522051.2 XP_011520353.1 3 - 4 305 94
XM_011522052.2 XP_011520354.1 2 - 3 363 94
XM_011522053.1 XP_011520355.1 2 - 3 144 94
XM_011522056.2 XP_011520358.2 2 - 3 530 94
XM_017022587.1 XP_016878076.1 2 - 3 530 94
XM_017022588.1 XP_016878077.1 2 - 3 530 94
XM_017022589.1 XP_016878078.1 2 - 3 530 94
XM_017022590.1 XP_016878079.1 3 - 4 282 94
XM_017022591.1 XP_016878080.1 2 - 3 229 94
XM_017022592.1 XP_016878081.1 2 - 3 376 94
XM_017022593.1 XP_016878082.1 2 - 3 530 94
XM_017022594.1 XP_016878083.1 1 - 2 65 94
XM_017022597.1 XP_016878086.1 2 - 3 530 94
XM_017022598.1 XP_016878087.1 2 - 3 530 94
XM_017022599.1 XP_016878088.1 2 - 3 530 94
GenBank mRNA AB075819.1 - 2 - 3 184 94
BC144297.1 - 2 - 3 161 94
BF898268.1 - 2 - 3 185 94

View Full Product Details