Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CLEC7A
- Entrez Gene ID
-
64581
- Gene Name
- C-type lectin domain family 7 member A
- Gene Aliases
-
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
- Chromosome Location
-
Chr.12: 10116777 - 10130269 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF400602.1 |
- |
3 - 4 |
258 |
83 |
View Full Product Details