Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
Chromosome Location
Chr.10: 115093365 - 115948999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207303.4 NP_997186.1 14 - 15 2614 72
XM_011539587.1 XP_011537889.1 14 - 15 2609 72
XM_011539588.2 XP_011537890.1 14 - 15 2159 72
XM_011539589.2 XP_011537891.1 14 - 15 2609 72
XM_011539590.2 XP_011537892.1 14 - 15 4511 72
XM_011539591.2 XP_011537893.1 14 - 15 2482 72
XM_017016035.1 XP_016871524.1 14 - 15 2609 72
XM_017016036.1 XP_016871525.1 14 - 15 2609 72
XM_017016037.1 XP_016871526.1 12 - 13 1613 72
XM_017016038.1 XP_016871527.1 13 - 14 9565 72
XM_017016039.1 XP_016871528.1 6 - 7 778 72
XM_017016040.1 XP_016871529.1 3 - 4 294 72
GenBank mRNA AB011106.2 - 8 - 9 1175 72
AK127277.1 - 14 - 15 2092 72
AY442317.1 - 14 - 15 2228 72

View Full Product Details