The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207303.4 | NP_997186.1 | 16 - 17 | 3018 | 61 |
XM_011539587.1 | XP_011537889.1 | 16 - 17 | 3013 | 61 | |
XM_011539588.2 | XP_011537890.1 | 16 - 17 | 2563 | 61 | |
XM_011539589.2 | XP_011537891.1 | 16 - 17 | 3013 | 61 | |
XM_011539590.2 | XP_011537892.1 | 16 - 17 | 4915 | 61 | |
XM_011539591.2 | XP_011537893.1 | 16 - 17 | 2886 | 61 | |
XM_017016035.1 | XP_016871524.1 | 16 - 17 | 3013 | 61 | |
XM_017016036.1 | XP_016871525.1 | 16 - 17 | 3013 | 61 | |
XM_017016037.1 | XP_016871526.1 | 14 - 15 | 2017 | 61 | |
XM_017016038.1 | XP_016871527.1 | 15 - 16 | 9969 | 61 | |
XM_017016039.1 | XP_016871528.1 | 8 - 9 | 1182 | 61 | |
XM_017016040.1 | XP_016871529.1 | 5 - 6 | 698 | 61 | |
GenBank mRNA | AB011106.2 | - | 10 - 11 | 1579 | 61 |
AK127277.1 | - | 16 - 17 | 2496 | 61 | |
AY442317.1 | - | 16 - 17 | 2632 | 61 |