The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276282.3 | NP_001263211.1 | 2 - 3 | 763 | 165 |
NM_207303.4 | NP_997186.1 | 2 - 3 | 763 | 165 | |
XM_011539587.1 | XP_011537889.1 | 2 - 3 | 758 | 165 | |
XM_011539589.2 | XP_011537891.1 | 2 - 3 | 758 | 165 | |
XM_017016035.1 | XP_016871524.1 | 2 - 3 | 758 | 165 | |
XM_017016036.1 | XP_016871525.1 | 2 - 3 | 758 | 165 | |
GenBank mRNA | AK127277.1 | - | 2 - 3 | 241 | 165 |
AY442317.1 | - | 2 - 3 | 377 | 165 | |
BC035157.1 | - | 2 - 3 | 769 | 165 | |
BC047716.1 | - | 2 - 3 | 708 | 165 | |
BC139916.1 | - | 2 - 3 | 752 | 165 | |
BC139923.1 | - | 2 - 3 | 752 | 165 |