Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
Chromosome Location
Chr.10: 115093365 - 115948999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276282.3 NP_001263211.1 6 - 7 1384 113
NM_207303.4 NP_997186.1 6 - 7 1384 113
NR_074088.2 8 - 9 1274 113
XM_011539587.1 XP_011537889.1 6 - 7 1379 113
XM_011539588.2 XP_011537890.1 6 - 7 929 113
XM_011539589.2 XP_011537891.1 6 - 7 1379 113
XM_011539590.2 XP_011537892.1 6 - 7 3281 113
XM_011539591.2 XP_011537893.1 6 - 7 1252 113
XM_017016035.1 XP_016871524.1 6 - 7 1379 113
XM_017016036.1 XP_016871525.1 6 - 7 1379 113
XM_017016037.1 XP_016871526.1 5 - 6 639 113
XM_017016038.1 XP_016871527.1 6 - 7 8591 113
GenBank mRNA AK127277.1 - 6 - 7 862 113
AY442317.1 - 6 - 7 998 113
BC035157.1 - 6 - 7 1390 113
BC047716.1 - 6 - 7 1329 113
BC139916.1 - 6 - 7 1373 113
BC139923.1 - 6 - 7 1373 113

View Full Product Details