The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017867.2 | NP_060337.2 | 5 - 6 | 715 | 66 |
XM_017008332.1 | XP_016863821.1 | 5 - 6 | 727 | 66 | |
GenBank mRNA | AK000541.1 | - | 5 - 6 | 670 | 66 |
AL136673.1 | - | 5 - 6 | 685 | 66 | |
BC010367.1 | - | 5 - 6 | 681 | 66 | |
BM905677.1 | - | 5 - 6 | 635 | 66 |