Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location
Chr.12: 98729904 - 99984727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152788.4 NP_690001.3 2 - 3 631 83
XM_005269028.4 XP_005269085.1 2 - 3 242 83
XM_005269029.4 XP_005269086.1 2 - 3 214 83
XM_006719504.3 XP_006719567.1 2 - 3 704 83
XM_006719505.3 XP_006719568.1 2 - 3 704 83
XM_006719506.3 XP_006719569.1 2 - 3 704 83
XM_006719507.3 XP_006719570.1 2 - 3 704 83
XM_006719508.3 XP_006719571.1 2 - 3 704 83
XM_006719509.3 XP_006719572.1 2 - 3 704 83
XM_006719510.3 XP_006719573.1 2 - 3 704 83
XM_006719512.3 XP_006719575.1 2 - 3 704 83
XM_006719513.3 XP_006719576.1 2 - 3 704 83
XM_006719514.3 XP_006719577.1 2 - 3 704 83
XM_011538571.2 XP_011536873.1 2 - 3 704 83
XM_017019651.1 XP_016875140.1 2 - 3 704 83
XM_017019652.1 XP_016875141.1 2 - 3 704 83
XM_017019653.1 XP_016875142.1 2 - 3 704 83
XM_017019654.1 XP_016875143.1 2 - 3 704 83
XM_017019655.1 XP_016875144.1 2 - 3 704 83
XM_017019656.1 XP_016875145.1 2 - 3 704 83
XM_017019657.1 XP_016875146.1 2 - 3 704 83
XM_017019658.1 XP_016875147.1 2 - 3 704 83
GenBank mRNA AK124453.1 - 2 - 3 377 83
AY281131.1 - 2 - 3 214 83

View Full Product Details