Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location
Chr.12: 98729904 - 99984727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152788.4 NP_690001.3 4 - 5 1084 59
XM_005269028.4 XP_005269085.1 4 - 5 695 59
XM_005269029.4 XP_005269086.1 4 - 5 667 59
XM_006719504.3 XP_006719567.1 4 - 5 1157 59
XM_006719505.3 XP_006719568.1 4 - 5 1157 59
XM_006719506.3 XP_006719569.1 4 - 5 1157 59
XM_006719507.3 XP_006719570.1 4 - 5 1157 59
XM_006719508.3 XP_006719571.1 4 - 5 1157 59
XM_006719509.3 XP_006719572.1 4 - 5 1157 59
XM_006719510.3 XP_006719573.1 4 - 5 1157 59
XM_006719512.3 XP_006719575.1 4 - 5 1157 59
XM_006719513.3 XP_006719576.1 4 - 5 1157 59
XM_006719514.3 XP_006719577.1 4 - 5 1157 59
XM_011538571.2 XP_011536873.1 4 - 5 1157 59
XM_017019651.1 XP_016875140.1 4 - 5 1157 59
XM_017019652.1 XP_016875141.1 4 - 5 1157 59
XM_017019653.1 XP_016875142.1 4 - 5 1157 59
XM_017019654.1 XP_016875143.1 4 - 5 1157 59
XM_017019655.1 XP_016875144.1 4 - 5 1157 59
XM_017019656.1 XP_016875145.1 4 - 5 1157 59
XM_017019657.1 XP_016875146.1 4 - 5 1157 59
XM_017019658.1 XP_016875147.1 4 - 5 1157 59
GenBank mRNA AK124453.1 - 4 - 5 830 59
AY281131.1 - 4 - 5 667 59

View Full Product Details