Product Details

Assay Reference Genome
Location

Chr.19:40395565 on build GRCh38
Cytoband
19q13.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
57716
Gene Symbol
PRX
Gene Name
periaxin
Gene Aliases
CMT4F
Location
Chr.19:40393762-40414718 on build GRCh38
Assay Gene Location
Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PRX NM_020956.2 6 3705 NP_066007.1
NM_181882.2 7 3056 NP_870998.2
XM_011527171.2 7 3048 XP_011525473.1
XM_017027046.1 3 3207 XP_016882535.1
XM_017027047.1 4 2750 XP_016882536.1
AB046840.1 3 3186 BAB13446.1
AF321191.1 7 3056 AAK19279.1
AF321192.1 6 3705 AAK19280.1
BC067266.1 7 3058 AAH67266.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833831 Chr19:40290833 - 40454958 on Build GRCh38 Loss C19orf47 SERTAD1 PLD3 BLVRB PRX LOC105372489 MIR6796 HIPK4 SERTAD3

View Full Product Details