The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040135.2 | NP_001035225.1 | 3 - 4 | 359 | 95 |
NM_020445.5 | NP_065178.1 | 3 - 4 | 359 | 95 | |
NR_073000.1 | 2 - 3 | 303 | 95 | ||
XM_006716065.2 | XP_006716128.1 | 3 - 4 | 341 | 95 | |
XM_006716066.1 | XP_006716129.1 | 3 - 4 | 341 | 95 | |
XM_006716067.3 | XP_006716130.1 | 3 - 4 | 341 | 95 | |
XM_011516425.2 | XP_011514727.1 | 3 - 4 | 341 | 95 | |
XM_011516426.2 | XP_011514728.1 | 3 - 4 | 341 | 95 | |
XM_011516427.1 | XP_011514729.1 | 3 - 4 | 341 | 95 | |
XM_017012460.1 | XP_016867949.1 | 3 - 4 | 341 | 95 | |
XM_017012461.1 | XP_016867950.1 | 3 - 4 | 341 | 95 | |
XM_017012462.1 | XP_016867951.1 | 2 - 3 | 233 | 95 | |
XM_017012463.1 | XP_016867952.1 | 2 - 3 | 172 | 95 | |
XM_017012464.1 | XP_016867953.1 | 2 - 3 | 300 | 95 | |
XM_017012465.1 | XP_016867954.1 | 1 - 2 | 126 | 95 | |
GenBank mRNA | AF023453.1 | - | 3 - 4 | 261 | 95 |
AF086920.1 | - | 2 - 3 | 222 | 95 | |
AK294292.1 | - | 2 - 3 | 257 | 95 | |
AK315534.1 | - | 3 - 4 | 341 | 95 | |
BC008682.1 | - | 3 - 4 | 314 | 95 | |
BC015207.1 | - | 3 - 4 | 271 | 95 | |
DN601600.1 | - | 3 - 4 | 359 | 95 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017012548.1 | XP_016868037.1 | 11 - 12 | 1382 | 95 |
XM_017012549.1 | XP_016868038.1 | 9 - 10 | 1302 | 95 | |
XM_017012550.1 | XP_016868039.1 | 8 - 9 | 997 | 95 |