The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145029.1 | NP_001138501.1 | 9 - 10 | 1508 | 99 |
XM_006722325.3 | XP_006722388.1 | 9 - 10 | 1336 | 99 | |
XM_011525662.2 | XP_011523964.1 | 9 - 10 | 1336 | 99 | |
XM_011525664.2 | XP_011523966.1 | 9 - 10 | 1336 | 99 | |
XM_011525665.2 | XP_011523967.1 | 9 - 10 | 1413 | 99 | |
XM_011525666.2 | XP_011523968.1 | 9 - 10 | 1413 | 99 | |
GenBank mRNA | AF269088.1 | - | 9 - 10 | 1509 | 99 |
BC028407.1 | - | - | 1416 | 99 |