The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101404.1 | NP_001094874.1 | 4 - 5 | 641 | 88 |
XM_011521895.2 | XP_011520197.1 | 4 - 5 | 1933 | 88 | |
XM_011521896.2 | XP_011520198.1 | 5 - 6 | 617 | 88 | |
XM_011521899.2 | XP_011520201.1 | 3 - 4 | 1408 | 88 | |
XM_017022487.1 | XP_016877976.1 | 5 - 6 | 850 | 88 | |
XM_017022488.1 | XP_016877977.1 | 3 - 4 | 425 | 88 |