Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C21orf62-AS1
Entrez Gene ID
54067
Gene Name
C21orf62 antisense RNA 1
Gene Aliases
C21orf49
Chromosome Location
Chr.21: 32772100 - 32797707 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024622.1 3 - 4 434 84
NR_024623.1 3 - 4 422 84
GenBank mRNA AK096601.1 - 3 - 4 422 84
BC117397.1 - 3 - 4 316 84
BC117399.1 - 3 - 4 328 84
DA776905.1 - 3 - 4 422 84


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CF272642.1 - 1 - 2 Chr. 21 - 32784895 - 32797318 [-] on Build GRCh38 84
GenBankmRNA BX477211.1 - 3 - 4 372 Chr. 21 - 32772162 - 32797168 [+] on Build GRCh38 84
GenBankmRNA AV717800.1 - 2 - 3 226 Chr. 21 - 32772174 - 32797446 [+] on Build GRCh38 84
GenBankmRNA AV655412.1 - 3 - 4 300 Chr. 21 - 32772235 - 32797082 [+] on Build GRCh38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream PAX3 and PAX7 binding protein 1 PAXBP1 BM020, C21orf66, FSAP105, GCFC, GCFC1 -16.8kb Sense Chr. - 32733899 - 32771858 on Build GRCh38
Downstream uncharacterized LOC102724502 LOC102724502 161.6kb Anti-sense Chr. - 32958888 - 32960566 on Build GRCh38

View Full Product Details