The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213602.2 | NP_998767.1 | 5 - 6 | 1056 | 74 |
GenBank mRNA | AK092746.1 | - | 2 - 3 | 607 | 74 |
AK095432.1 | - | 6 - 7 | 760 | 74 | |
AK172835.1 | - | 5 - 6 | 1056 | 74 | |
AK290400.1 | - | 5 - 6 | 910 | 74 | |
AK290402.1 | - | 5 - 6 | 920 | 74 | |
AK301189.1 | - | 5 - 6 | 937 | 74 | |
AY971516.1 | - | 5 - 6 | 1045 | 74 |