The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206827.1 | NP_996563.1 | 3 - 4 | 880 | 87 |
GenBank mRNA | AY439004.1 | - | 3 - 4 | 880 | 87 |
BC132703.1 | - | 3 - 4 | 616 | 87 | |
BC136761.1 | - | 3 - 4 | 616 | 87 |