Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK097913.1 |
- |
1 - 2 |
144 |
|
Chr. 14 - 22226674 - 22228439 [+] on Build GRCh38 |
76 |
GenBankmRNA |
HY077901.1 |
- |
1 - 2 |
102 |
|
Chr. 14 - 22226717 - 22547522 [+] on Build GRCh38 |
76 |
GenBankmRNA |
DB108367.1 |
- |
1 - 2 |
145 |
|
Chr. 14 - 22226674 - 22227401 [+] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
T cell receptor delta variable 1 |
TRDV1 |
hDV101S1 |
-130.1kb
|
Sense |
Chr. - 22096050 - 22096619 on Build GRCh38 |
Downstream |
T cell receptor delta variable 2 |
TRDV2 |
hDV102S1 |
194.1kb
|
Sense |
Chr. - 22422546 - 22423042 on Build GRCh38 |
View Full Product Details