The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019597.4 | NP_062543.1 | 1 - 2 | 191 | 61 |
GenBank mRNA | AK298133.1 | - | 1 - 2 | 98 | 61 |
DA686966.1 | - | 1 - 2 | 191 | 61 |