Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
Chromosome Location
Chr.3: 8619400 - 8745035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006530.1 XP_016862019.1 1 - 2 188 95
GenBank mRNA AK093066.1 - 1 - 2 189 95
BC043244.1 - 1 - 2 197 95

View Full Product Details