Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A10
Entrez Gene ID
387775
Gene Name
solute carrier family 22 member 10
Gene Aliases
OAT5, hOAT5
Chromosome Location
Chr.11: 63268257 - 63311983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039752.3 NP_001034841.3 6 - 7 1271 88
NR_134874.1 7 - 8 1026 88
XM_011545016.2 XP_011543318.1 8 - 9 2276 88
XM_017017699.1 XP_016873188.1 7 - 8 1073 88
XM_017017700.1 XP_016873189.1 6 - 7 1509 88
XM_017017701.1 XP_016873190.1 8 - 9 2276 88
XM_017017703.1 XP_016873192.1 5 - 6 617 88
XM_017017704.1 XP_016873193.1 6 - 7 1508 88
GenBank mRNA AB075876.1 - 7 - 8 1026 88
AK298430.1 - 7 - 8 1073 88
BC171744.1 - 4 - 5 606 88

View Full Product Details