The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039752.3 | NP_001034841.3 | 7 - 8 | 1490 | 77 |
XM_011545016.2 | XP_011543318.1 | 9 - 10 | 2495 | 77 | |
XM_017017699.1 | XP_016873188.1 | 8 - 9 | 1292 | 77 | |
XM_017017700.1 | XP_016873189.1 | 7 - 8 | 1728 | 77 | |
XM_017017703.1 | XP_016873192.1 | 6 - 7 | 836 | 77 |