Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCU
Entrez Gene ID
90550
Gene Name
mitochondrial calcium uniporter
Gene Aliases
C10orf42, CCDC109A
Chromosome Location
Chr.10: 72692131 - 72887694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270679.1 NP_001257608.1 2 - 3 236 94
NM_001270680.1 NP_001257609.1 2 - 3 513 94
NM_138357.2 NP_612366.1 2 - 3 236 94
NR_073062.1 4 - 5 401 94
XM_017016882.1 XP_016872371.1 2 - 3 436 94
GenBank mRNA AK075439.1 - 2 - 3 224 94
AK128016.1 - 2 - 3 513 94
AK301475.1 - 4 - 5 382 94
AK310455.1 - 5 - 6 406 94
AK311133.1 - 4 - 5 395 94
AK315519.1 - 2 - 3 219 94
BC010682.1 - 2 - 3 193 94
BC034235.1 - 2 - 3 236 94
HY146050.1 - 2 - 3 238 94

View Full Product Details