The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270679.1 | NP_001257608.1 | 7 - 8 | 931 | 153 |
NM_001270680.1 | NP_001257609.1 | 7 - 8 | 1271 | 153 | |
NM_138357.2 | NP_612366.1 | 7 - 8 | 994 | 153 | |
NR_073062.1 | 9 - 10 | 1159 | 153 | ||
XM_017016882.1 | XP_016872371.1 | 7 - 8 | 1194 | 153 | |
GenBank mRNA | AK025699.1 | - | 3 - 4 | 336 | 153 |
AK075439.1 | - | 7 - 8 | 981 | 153 | |
AK128016.1 | - | 7 - 8 | 1271 | 153 | |
AK315519.1 | - | 7 - 8 | 977 | 153 | |
BC010682.1 | - | 7 - 8 | 888 | 153 | |
BC034235.1 | - | 7 - 8 | 994 | 153 |