The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006600.3 | NP_006591.1 | 1 - 2 | 210 | 88 |
XM_017000095.1 | XP_016855584.1 | 1 - 2 | 209 | 88 | |
GenBank mRNA | AB019408.1 | - | 1 - 2 | 157 | 88 |
AF086922.1 | - | 1 - 2 | 173 | 88 | |
AF100760.1 | - | 1 - 2 | 173 | 88 | |
AF125465.1 | - | 1 - 2 | 181 | 88 | |
AF130736.1 | - | 1 - 2 | 124 | 88 | |
AL136725.1 | - | 1 - 2 | 185 | 88 | |
BC002399.2 | - | 1 - 2 | 122 | 88 | |
BC003132.1 | - | 1 - 2 | 130 | 88 | |
BC006147.1 | - | 1 - 2 | 156 | 88 | |
BC007280.1 | - | 1 - 2 | 199 | 88 | |
BC015153.1 | - | 1 - 2 | 198 | 88 | |
BC021139.1 | - | 1 - 2 | 171 | 88 | |
BP348144.1 | - | 1 - 2 | 210 | 88 |