Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM2
Entrez Gene ID
7750
Gene Name
zinc finger MYM-type containing 2
Gene Aliases
FIM, MYM, RAMP, SCLL, ZNF198
Chromosome Location
Chr.13: 19957412 - 20091845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190964.2 NP_001177893.1 10 - 11 2041 87
NM_001190965.2 NP_001177894.1 9 - 10 2056 87
NM_003453.4 NP_003444.1 10 - 11 2149 87
NM_197968.3 NP_932072.1 9 - 10 2102 87
XM_005266520.3 XP_005266577.1 9 - 10 2061 87
XM_011535216.2 XP_011533518.1 10 - 11 1731 87
XM_011535222.1 XP_011533524.1 9 - 10 1992 87
XM_011535223.2 XP_011533525.1 3 - 4 4446 87
XM_017020728.1 XP_016876217.1 9 - 10 1992 87
XM_017020729.1 XP_016876218.1 9 - 10 1967 87
XM_017020731.1 XP_016876220.1 10 - 11 1784 87
XM_017020732.1 XP_016876221.1 10 - 11 1806 87
XM_017020733.1 XP_016876222.1 11 - 12 1833 87
XM_017020734.1 XP_016876223.1 11 - 12 1819 87
XM_017020735.1 XP_016876224.1 10 - 11 1783 87
XM_017020736.1 XP_016876225.1 10 - 11 1955 87
XM_017020737.1 XP_016876226.1 10 - 11 1730 87
XM_017020738.1 XP_016876227.1 9 - 10 2045 87
GenBank mRNA AF035374.1 - 7 - 8 1347 87
AF060181.1 - 7 - 8 1399 87
AJ224901.1 - 10 - 11 2035 87
AK226118.1 - 9 - 10 2036 87
AK289741.1 - 9 - 10 2027 87
AK302917.1 - 4 - 5 416 87
BC036372.1 - 9 - 10 2102 87
BX647944.1 - 9 - 10 2056 87
BX648905.1 - 2 - 3 2486 87
Y13472.1 - 10 - 11 2047 87

View Full Product Details