The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190964.2 | NP_001177893.1 | 15 - 16 | 2681 | 147 |
NM_001190965.2 | NP_001177894.1 | 14 - 15 | 2696 | 147 | |
NM_003453.4 | NP_003444.1 | 15 - 16 | 2789 | 147 | |
NM_197968.3 | NP_932072.1 | 14 - 15 | 2742 | 147 | |
XM_005266520.3 | XP_005266577.1 | 14 - 15 | 2701 | 147 | |
XM_011535216.2 | XP_011533518.1 | 15 - 16 | 2371 | 147 | |
XM_011535222.1 | XP_011533524.1 | 14 - 15 | 2632 | 147 | |
XM_011535223.2 | XP_011533525.1 | 8 - 9 | 5086 | 147 | |
XM_011535224.2 | XP_011533526.1 | 8 - 9 | 987 | 147 | |
XM_017020728.1 | XP_016876217.1 | 14 - 15 | 2632 | 147 | |
XM_017020729.1 | XP_016876218.1 | 14 - 15 | 2607 | 147 | |
XM_017020731.1 | XP_016876220.1 | 15 - 16 | 2424 | 147 | |
XM_017020732.1 | XP_016876221.1 | 15 - 16 | 2446 | 147 | |
XM_017020733.1 | XP_016876222.1 | 16 - 17 | 2473 | 147 | |
XM_017020734.1 | XP_016876223.1 | 16 - 17 | 2459 | 147 | |
XM_017020735.1 | XP_016876224.1 | 15 - 16 | 2423 | 147 | |
XM_017020736.1 | XP_016876225.1 | 15 - 16 | 2595 | 147 | |
XM_017020737.1 | XP_016876226.1 | 15 - 16 | 2370 | 147 | |
XM_017020738.1 | XP_016876227.1 | 14 - 15 | 2685 | 147 | |
GenBank mRNA | AF012126.1 | - | 8 - 9 | 1084 | 147 |
AF035374.1 | - | 12 - 13 | 1987 | 147 | |
AF060181.1 | - | 12 - 13 | 2039 | 147 | |
AJ224901.1 | - | 15 - 16 | 2675 | 147 | |
AK226118.1 | - | 14 - 15 | 2676 | 147 | |
AK289741.1 | - | 14 - 15 | 2667 | 147 | |
AK302917.1 | - | 9 - 10 | 1056 | 147 | |
BC036372.1 | - | 14 - 15 | 2742 | 147 | |
BX647944.1 | - | 14 - 15 | 2696 | 147 | |
BX648905.1 | - | 7 - 8 | 3126 | 147 | |
Y13472.1 | - | 15 - 16 | 2687 | 147 |